Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trioctylamine

Catalog No. T81000500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T81000500G 500 g
T8100025KG 2.5 kg
T81000100G 100 g
T8100025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. T81000500G Supplier Sigma Aldrich Supplier No. T81000500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trioctylamine is used as an extractant for organic acids (such as TCA, succinic acid, and acetic acid), and precious metals.

Specifications

CAS 1116-76-3
Density 0.809 g/mL (at 25°C (literature))
Boiling Point 164°C to 168°C (0.7 mmHg), 365°C to 367°C (lit.)
Molecular Formula C24H51N
Refractive Index n20/D 1.449 (literature)
Linear Formula [CH3(CH2)7]3 N
MDL Number MFCD00009560
Quantity 500 g
Synonym TOA
Molecular Weight (g/mol) 353.67
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.