Learn More
Medchemexpress LLC Trioctylamine-d6 | 1219798-90-9 | 99.1% | 359.71 | 1 MG
SDP
Catalog No. 5000452882
Encompass_Preferred
Trioctylamine-d6 is the deuterium labeled Trioctylamine. It is used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-52882 Supplier Medchemexpress LLC Supplier No. HYW011133S1MG
Trioctylamine-d6 is the deuterium labeled Trioctylamine. It is used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs