Learn More
Medchemexpress LLC Trioctylamine-d6 | 1219798-90-9 | 99.1% | 359.71 | 10 MG
SDP
Catalog No. 5000451807
Encompass_Preferred
Trioctylamine-d6 is the deuterium labeled Trioctylamine. It is used for research purposes and is not sold to patients. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during the drug development process.
Catalog No. 50-004-51807 Supplier Medchemexpress LLC Supplier No. HYW011133S10MG
Trioctylamine-d6 is the deuterium labeled Trioctylamine. It is used for research purposes and is not sold to patients. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during the drug development process.