Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trioctylphosphine

Catalog No. 718165500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-816-5500ML 500 mL
71-816-5100ML 100 mL
71-816-52L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 71-816-5500ML Supplier Sigma Aldrich Supplier No. 718165500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trioctylphosphine can be used for the conversion of metal nanocrystals, bulk powders, foils, wires, thin films to metal phosphides. It can also act as a solvent and stabilizer for synthesizing cadmium sulfide nanorods from cadmium acetate and sulfur.

Specifications

CAS 4731-53-7
Density 0.831 g/mL at 25°C
Boiling Point 284°C to 291°C (50 mmHg, lit.)
Molecular Formula C24H51P
Refractive Index n20/D 1.468
Linear Formula [CH3(CH2)7]3P
MDL Number MFCD00015298
Quantity 500 mL
Synonym TOP; P(Oct)3
Molecular Weight (g/mol) 370.64 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.