Learn More
Strem, An Ascensus Company CAS# 78-50-2. 25g. Trioctylphosphine oxide, 99% TOPO. MFCD00002083
SDP
Catalog No. 5090110461
Encompass
CAS# 78-50-2. 25g. Trioctylphosphine oxide, 99% TOPO. MFCD00002083. Molecular Weight: 386.65. Molecular Formula: (n-C8H17)3PO. Color/form: white to off-white solid Strem# 15-6661. http://www.strem.com/catalog/v/15-6661/
Catalog No. 50-901-10461 Supplier Strem, An Ascensus Company Supplier No. 15666125G
CAS# 78-50-2. 25g. Trioctylphosphine oxide, 99% TOPO. MFCD00002083. Molecular Weight: 386.65. Molecular Formula: (n-C8H17)3PO. Color/form: white to off-white solid Strem# 15-6661. http://www.strem.com/catalog/v/15-6661/