Learn More
Strem, An Ascensus Company CAS# 78-50-2. 100g. Trioctylphosphine oxide, min. 90% TOPO. MFCD00002083
SDP
Catalog No. 5090110459
Encompass
CAS# 78-50-2. 100g. Trioctylphosphine oxide, min. 90% TOPO. MFCD00002083. Molecular Weight: 386.65. Molecular Formula: (n-C8H17)3PO. Color/form: off-white xtl. Strem# 15-6660. http://www.strem.com/catalog/v/15-6660/
Catalog No. 50-901-10459 Supplier Strem, An Ascensus Company Supplier No. 156660100G
CAS# 78-50-2. 100g. Trioctylphosphine oxide, min. 90% TOPO. MFCD00002083. Molecular Weight: 386.65. Molecular Formula: (n-C8H17)3PO. Color/form: off-white xtl. Strem# 15-6660. http://www.strem.com/catalog/v/15-6660/