Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trioctylpropylammonium Chloride Pract., 95%, Thermo Scientific™

Catalog No. AC422140250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC422140250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC422140250 Supplier Thermo Scientific Chemicals Supplier No. 422140250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40739-43-3
Molecular Formula C27H58ClN
Molecular Weight (g/mol) 432.218
InChI Key WFEXFNMTEBFLMM-UHFFFAOYSA-M
Synonym 1-octanaminium, n,n-dioctyl-n-propyl-, chloride, trioctylpropylammonium chloride, 1-octanaminium, n,n-dioctyl-n-propyl-, chloride 1:1, trioctylpropylammoniumchloride, trioctyl propyl azanium chloride, tri-n-octylpropylammonium chloride, trioctylpropylammonium chloride pract, 1-octanaminium,n,n-dioctyl-n-propyl-, chloride 1:1
PubChem CID 2735155
IUPAC Name trioctyl(propyl)azanium;chloride
SMILES CCCCCCCC[N+](CCC)(CCCCCCCC)CCCCCCCC.[Cl-]

Specifications

Color Colorless
Infrared Spectrum Authentic
Linear Formula (CH3(CH2)7)3N(Cl)CH2CH2CH3
Quantity 25 g
Formula Weight 432.21
Percent Purity ≥94%
Physical Form Viscous Liquid After Melting
Chemical Name or Material Trioctylpropylammonium chloride, pract., 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.