Learn More
Medchemexpress LLC Triolein | 122-32-7 | MFCD00137563 | 885.43 | 1 G
SDP
Catalog No. 5000415682
Encompass_Preferred
Triolein is a symmetric triacylglycerol that reduces upregulation of MMP-1 and possesses strong antioxidant and anti-inflammatory activities.
  • Reduces upregulation of MMP-1.
  • Possesses strong antioxidant activity.
  • Possesses strong anti-inflammatory activity.
  • Available in deuterium-labeled versions.
  • Available in 13C-labeled versions.
  • Available as an analytical standard.
Catalog No. 50-004-15682 Supplier Medchemexpress LLC Supplier No. HYN19811G
Triolein is a symmetric triacylglycerol that reduces upregulation of MMP-1 and possesses strong antioxidant and anti-inflammatory activities.
  • Reduces upregulation of MMP-1.
  • Possesses strong antioxidant activity.
  • Possesses strong anti-inflammatory activity.
  • Available in deuterium-labeled versions.
  • Available in 13C-labeled versions.
  • Available as an analytical standard.