Learn More
Medchemexpress LLC Triolein | 122-32-7 | MFCD00137563 | 885.43 | 100 MG
SDP
Catalog No. 5000416029
Encompass_Preferred
Triolein (Standard) is the analytical standard of Triolein, intended for research and analytical applications. It is a symmetric triacylglycerol that reduces upregulation of MMP-1 and possesses strong antioxidant and anti-inflammatory activities.
  • Analytical standard for Triolein.
  • Intended for research and analytical applications.
  • Symmetric triacylglycerol.
  • Reduces upregulation of MMP-1.
  • Has strong antioxidant and anti-inflammatory activities.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-16029 Supplier Medchemexpress LLC Supplier No. HYN1981R100MG
Triolein (Standard) is the analytical standard of Triolein, intended for research and analytical applications. It is a symmetric triacylglycerol that reduces upregulation of MMP-1 and possesses strong antioxidant and anti-inflammatory activities.
  • Analytical standard for Triolein.
  • Intended for research and analytical applications.
  • Symmetric triacylglycerol.
  • Reduces upregulation of MMP-1.
  • Has strong antioxidant and anti-inflammatory activities.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.