Learn More
Medchemexpress LLC Triolein | 122-32-7 | MFCD00137563 | 99.9% | 885.43 | C57H104O6 | 10 MG
SDP
Catalog No. 5000295704
Encompass_Preferred
Triolein analytical standard is a symmetric triacylglycerol used as a reference standard in lipid analysis and research. It has molecular formula C57H104O6, molecular weight 885.43, CAS 122-32-7, and is supplied as a high-purity (99.88%) colorless to light yellow liquid for analytical applications.
  • High purity for reliable analytical results.
  • Liquid reference standard with consistent appearance.
  • Suitable for method development, calibration, and quality control in lipid analysis.
  • Offered in small quantity packaging for trace-level studies.
  • Includes definitive identifiers (molecular formula C57H104O6 and CAS 122-32-7) for unambiguous identification.
Catalog No. 50-002-95704 Supplier Medchemexpress LLC Supplier No. HYN1981R10MG
Triolein analytical standard is a symmetric triacylglycerol used as a reference standard in lipid analysis and research. It has molecular formula C57H104O6, molecular weight 885.43, CAS 122-32-7, and is supplied as a high-purity (99.88%) colorless to light yellow liquid for analytical applications.
  • High purity for reliable analytical results.
  • Liquid reference standard with consistent appearance.
  • Suitable for method development, calibration, and quality control in lipid analysis.
  • Offered in small quantity packaging for trace-level studies.
  • Includes definitive identifiers (molecular formula C57H104O6 and CAS 122-32-7) for unambiguous identification.