Learn More
Selleck Chemical LLC Triolein S3590-25mg
SDP
Catalog No. 503109881
Encompass_Preferred
Triolein is an inhibitor of metalloproteinase-1 (MMP-1) and reduces both the expression of IL-6 and ROS generation in irradiated keratinocytes
Catalog No. 50-310-9881 Supplier Selleck Chemical LLC Supplier No. S359025MG
Triolein is an inhibitor of metalloproteinase-1 (MMP-1) and reduces both the expression of IL-6 and ROS generation in irradiated keratinocytes