Learn More
Selleck Chemical LLC Triolein S3590-1g
SDP
Catalog No. 503110150
Encompass_Preferred
Triolein is an inhibitor of metalloproteinase-1 (MMP-1) and reduces both the expression of IL-6 and ROS generation in irradiated keratinocytes
Catalog No. 50-311-0150 Supplier Selleck Chemical LLC Supplier No. S35901G
Triolein is an inhibitor of metalloproteinase-1 (MMP-1) and reduces both the expression of IL-6 and ROS generation in irradiated keratinocytes