Learn More
Medchemexpress LLC Triolein-d5 | 60892-83-3 | 890.46 | 5 MG
SDP
Catalog No. 5000415883
Encompass_Preferred
Triolein-d5 is the deuterium labeled Triolein. Triolein is a symmetrical triacylglycerol that reduces MMP-1 upregulation, with strong antioxidant and anti-inflammatory properties. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-15883 Supplier Medchemexpress LLC Supplier No. HYN1981S15MG
Triolein-d5 is the deuterium labeled Triolein. Triolein is a symmetrical triacylglycerol that reduces MMP-1 upregulation, with strong antioxidant and anti-inflammatory properties. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS