Learn More
American Radiolabeled Chemicals Inc TRIOLEIN-GLY-14C(U) 250UC
SDP
Catalog No. 5090104116
Encompass_Preferred
Triolein-glycerol-14C(U) 250uCi in toluene:ethanol (1:1), 0.1 mCi/ml, S.A. 50-100 mCi/mmol, M.W. 885.4, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-04116 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 3076250UC
Add to Cart
Add to Cart
Request a Quote
Triolein-glycerol-14C(U) 250uCi in toluene:ethanol (1:1), 0.1 mCi/ml, S.A. 50-100 mCi/mmol, M.W. 885.4, Shipped in Dry Ice, Exclusive