Learn More
Medchemexpress LLC Triolein (standard) | 122-32-7 | 885.43 | 25 MG
SDP
Catalog No. 5000389274
Encompass_Preferred
Triolein (Standard) is the analytical standard of Triolein intended for research and analytical applications. It is a symmetric triacylglycerol that reduces upregulation of MMP-1 and has strong antioxidant and anti-inflammatory activities. This compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS, serving as a reference standard supplied for assay.
  • Analytical standard for research
  • Symmetric triacylglycerol
  • Reduces upregulation of MMP-1
  • Strong antioxidant and anti-inflammatory activities
  • Suitable for HPLC research experiments
  • Suitable for GC research experiments
  • Suitable for MS research experiments
Catalog No. 50-003-89274 Supplier Medchemexpress LLC Supplier No. 25MGHYN1981R
Triolein (Standard) is the analytical standard of Triolein intended for research and analytical applications. It is a symmetric triacylglycerol that reduces upregulation of MMP-1 and has strong antioxidant and anti-inflammatory activities. This compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS, serving as a reference standard supplied for assay.
  • Analytical standard for research
  • Symmetric triacylglycerol
  • Reduces upregulation of MMP-1
  • Strong antioxidant and anti-inflammatory activities
  • Suitable for HPLC research experiments
  • Suitable for GC research experiments
  • Suitable for MS research experiments