Learn More
Pickering Laboratories Inc TRIONE REAGENT 4X950ML
SDP
Catalog No. NC9438300
Encompass_Preferred
TRIONE Ninhydrin Reagent (4-month* shelf life), case of 4 (950 mL/bottle) Post-column Reagent. Amino Acid Analysis.
Catalog No. NC9438300 Supplier Pickering Laboratories Inc Supplier No. T100C
TRIONE Ninhydrin Reagent (4-month* shelf life), case of 4 (950 mL/bottle) Post-column Reagent. Amino Acid Analysis.