Learn More
TA Instruments TRIOS Guardian, Software for 21 CFR Part 11 Compliance, Restricted Access, User Levels, Audit Trail, Results Log, PDF Creation, File Integrity Check
Catalog No. 5000580595 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments TRIOS Guardian is designed primarily for laboratories in regulated environments. TRIOS Guardian provides software tools that aid with 21 CFR Part 11 compliance. It allows users to restrict instrument control access, set defined user levels, and record analysis events. Additionally, TRIOS Guardian provides options to define access rights, restrict calibration and method development, generate audit trails, generate results logs, create PDF files, and perform file integrity checks.
Catalog No. 50-005-80595 Supplier TA Instruments Supplier No. 926020.001
Waters TA Instruments TRIOS Guardian is designed primarily for laboratories in regulated environments. TRIOS Guardian provides software tools that aid with 21 CFR Part 11 compliance. It allows users to restrict instrument control access, set defined user levels, and record analysis events. Additionally, TRIOS Guardian provides options to define access rights, restrict calibration and method development, generate audit trails, generate results logs, create PDF files, and perform file integrity checks.