Learn More
TA Instruments TRIOS Kinetics Analysis Suite, Advanced Analysis Tool for DSC, TGA, SDT, Follows ASTM Standards, Calculates Activation Energy, Reaction Order, Half Life, Compatible with Thermal Analyzers
Catalog No. 5000580670 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments TRIOS Kinetics Analysis Suite is a powerful tool for the specialized analysis of data obtained from Waters TA Instruments thermal analyzers including DSC, TGA, and SDT. Following ASTM standards (E537, E698, E1231, E1641, E2041, and E2070), this premium feature enables analysis of time and temperature-dependent processes. It determines parameters such as activation energy (Ea), reaction order (n), and half-life (t1/2), helping users make informed predictions about material behavior under various experimental conditions.
Catalog No. 50-005-80670 Supplier TA Instruments Supplier No. 926022.001
Waters TA Instruments TRIOS Kinetics Analysis Suite is a powerful tool for the specialized analysis of data obtained from Waters TA Instruments thermal analyzers including DSC, TGA, and SDT. Following ASTM standards (E537, E698, E1231, E1641, E2041, and E2070), this premium feature enables analysis of time and temperature-dependent processes. It determines parameters such as activation energy (Ea), reaction order (n), and half-life (t1/2), helping users make informed predictions about material behavior under various experimental conditions.