Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Triosephosphate isomerase (TPI) | 9023-78-3 | 1 MG
SDP

Catalog No. 5000398437
Encompass_Preferred

Triosephosphate isomerase (TPI) is a glycolytic enzyme that rapidly interconverts dihydroxyacetone phosphate and D-glyceraldehyde-3-phosphate. Its catalytic site is located at the dimer interface. This product is for research use only.

  • Glycolytic enzyme
  • Interconverts dihydroxyacetone phosphate and D-glyceraldehyde-3-phosphate
  • Catalytic site at the dimer interface
  • Solid appearance
  • White to off-white color
  • Specific activity: ≥10000 U/mg protein
  • EC number: 5.3.1.1

Catalog No. 50-003-98437 Supplier Medchemexpress LLC Supplier No. HYP29311MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triosephosphate isomerase (TPI) is a glycolytic enzyme that rapidly interconverts dihydroxyacetone phosphate and D-glyceraldehyde-3-phosphate. Its catalytic site is located at the dimer interface. This product is for research use only.

  • Glycolytic enzyme
  • Interconverts dihydroxyacetone phosphate and D-glyceraldehyde-3-phosphate
  • Catalytic site at the dimer interface
  • Solid appearance
  • White to off-white color
  • Specific activity: ≥10000 U/mg protein
  • EC number: 5.3.1.1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.