Learn More
Medchemexpress LLC Triosephosphate isomerase (TPI) | 9023-78-3 | 5 MG
SDP
Catalog No. 5000396603
Encompass_Preferred
Triosephosphate isomerase (TPI) is a glycolytic enzyme that rapidly interconverts dihydroxyacetone phosphate and D-glyceraldehyde-3-phosphate. Its catalytic site is located at the dimer interface. This product is intended for research use only.
  • Specific activity is ≥10000 U/mg protein.
  • One unit is defined as the amount of enzyme that will convert 1.0 μmole D-glyceraldehyde 3-phosphate to dihydroxyacetone phosphate per minute at pH 7.6 at 25°C.
  • Available in solid form.
  • Appears as white to off-white in color.
  • Shipping at room temperature in continental US; may vary elsewhere.
Catalog No. 50-003-96603 Supplier Medchemexpress LLC Supplier No. HYP29315MG
Triosephosphate isomerase (TPI) is a glycolytic enzyme that rapidly interconverts dihydroxyacetone phosphate and D-glyceraldehyde-3-phosphate. Its catalytic site is located at the dimer interface. This product is intended for research use only.
  • Specific activity is ≥10000 U/mg protein.
  • One unit is defined as the amount of enzyme that will convert 1.0 μmole D-glyceraldehyde 3-phosphate to dihydroxyacetone phosphate per minute at pH 7.6 at 25°C.
  • Available in solid form.
  • Appears as white to off-white in color.
  • Shipping at room temperature in continental US; may vary elsewhere.