Learn More
Enzo Life Sciences TRIP-Br1 monoclonal antibody (TB1-27/2) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804645C100
Encompass_Preferred
Clone: TB1-27/2. Isotype: IgG2a. Immunogen: Synthetic peptide corresponding to aa 27-42 (D27PGHAAVAQAPPAVAS42) of human TRIP-Br1 [SEI-1]. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse Porcine, Monkey. Applications: IHC, IP, WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9964