Learn More
Aviva Systems Biology Corp TRIP10 Antibody - middle region (ARP38438_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38438_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIP10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIP10. Alternate names: CIP4, HSTP, STOT, STPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56657