Learn More
Aviva Systems Biology Corp TRIP13 Antibody - C-terminal region (ARP38434_T100) | 100uL
SDP
Catalog No. 5001456656
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIP13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRIP13. Alternate names: 16E1BPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56656 Supplier Aviva Systems Biology Corp Supplier No. ARP38434_T100
This is a rabbit polyclonal antibody against TRIP13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRIP13. Alternate names: 16E1BPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat