Learn More
Aviva Systems Biology Corp TRIP13 Antibody - N-terminal region (ARP38433_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38433_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIP13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIP13. Alternate names: 16E1BPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53855