Learn More
Aviva Systems Biology Corp TRIP6 Antibody - middle region (ARP51618_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51618_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIP6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIP6. Alternate names: MGC10556, MGC10558, MGC29959, MGC3837, MGC4423, OIP1, ZRP-1, OIP-1, TRIP-6, TRIP6i2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60571