Learn More
Medchemexpress LLC Tripelennamine (hydrochloride) | 154-69-8 | 99.8% | 291.82 | 1 ML
SDP
Catalog No. 5000386196
Encompass_Preferred
Tripelennamine hydrochloride is an ethylenediamine derivative that acts as a potent histamine H1-receptor antagonist. It is used to reduce allergic responses caused by histamine and can be researched for conditions such as rhinitis, conjunctivitis, and allergic and anaphylactic reactions. It is for research use only and not sold to patients.
  • Potent histamine H1-receptor antagonist
  • Ethylenediamine derivative
  • For research of rhinitis, conjunctivitis, and allergic and anaphylactic reactions
  • For research use only
  • 99.81% purity
Catalog No. 50-003-86196 Supplier Medchemexpress LLC Supplier No. HY1742810MM/1ML
Tripelennamine hydrochloride is an ethylenediamine derivative that acts as a potent histamine H1-receptor antagonist. It is used to reduce allergic responses caused by histamine and can be researched for conditions such as rhinitis, conjunctivitis, and allergic and anaphylactic reactions. It is for research use only and not sold to patients.
  • Potent histamine H1-receptor antagonist
  • Ethylenediamine derivative
  • For research of rhinitis, conjunctivitis, and allergic and anaphylactic reactions
  • For research use only
  • 99.81% purity