Learn More
Medchemexpress LLC Tripelennamine hydrochloride | 154-69-8 | 99.8% | 291.82 | 250 MG
SDP
Catalog No. 5000384433
Encompass_Preferred
Tripelennamine hydrochloride is an ethylenediamine derivative and a potent histamine H1-receptor antagonist. It lessens the allergic response of the organism caused by histamine and can be used for the research of rhinitis, conjunctivitis, and allergic and anaphylactic reactions.
  • Potent histamine H1-receptor antagonist
  • Lessens allergic response
  • Used for research of rhinitis, conjunctivitis, and allergic and anaphylactic reactions
Catalog No. 50-003-84433 Supplier Medchemexpress LLC Supplier No. 250MGHY17428
Tripelennamine hydrochloride is an ethylenediamine derivative and a potent histamine H1-receptor antagonist. It lessens the allergic response of the organism caused by histamine and can be used for the research of rhinitis, conjunctivitis, and allergic and anaphylactic reactions.
  • Potent histamine H1-receptor antagonist
  • Lessens allergic response
  • Used for research of rhinitis, conjunctivitis, and allergic and anaphylactic reactions