Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tripentylamine

Catalog No. 472034250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-203-4250ML 250 mL
47-203-42L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-203-4250ML Supplier Sigma Aldrich Supplier No. 472034250ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The product is an isomeric mixture of tripentylamine (tpa)
  • Tpa is a tertiary amine
  • It forms charge transfer complexes with c60 in toluene
  • The stability constants of these complexes have been evaluated using the benesi-hildebrand equation

Specifications

CAS 621-77-2
Density 0.782 g/mL (at 25°C (literature))
Boiling Point 81°C to 83°C (0.2 mmHg)
Molecular Formula C15H33N
Refractive Index n20/D 1.436 (literature)
Linear Formula [CH3(CH2)4]3 N
MDL Number MFCD00015270
Quantity 250 mL
Synonym Triamylamine, mixed isomers
Molecular Weight (g/mol) 227.43
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.