Learn More
Medchemexpress LLC Tripeptide-32 | 1185583-20-3 | 99.95% | 302.33 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP52445MG
Encompass_Preferred
Tripeptide-32 is a bioactive peptide that targets the PER1 gene. It can regulate circadian rhythms and stimulate the synthesis of cellular repair proteins, exhibiting anti-aging activity.
  • Targets the PER1 gene
  • Regulates circadian rhythms
  • Stimulates synthesis of cellular repair proteins
  • Exhibits anti-aging activity
  • For research use only
  • Available as a solid
  • White to off-white color
  • Soluble in DMSO at 125 mg/mL (413.46 mM), requiring ultrasonic
Catalog No. 50-003-99646