Learn More
Medchemexpress LLC Tripeptide epoxyketone immunoproteasome (LMP-7) inhibitor | 1416709-79-9 | 98.9% | 618.72 g/mol | C37H38N4O5 | 50MG
SDP
Catalog No. 5000239468
Encompass_Preferred
A selective tripeptide epoxyketone inhibitor of the immunoproteasome LMP7 subunit for biochemical and cellular research. It covalently modifies proteasomal N-terminal threonine active sites and has demonstrated antitumor activity in preclinical studies.
  • Selective LMP7 (β5i) immunoproteasome inhibitor.
  • Covalently modifies N-terminal threonine active sites.
  • Reported IC50 of 22 nM against LMP7.
  • Demonstrated inhibition of multiple myeloma cell growth and induction of apoptosis.
  • High chemical purity (98.9%).
  • Enantiomeric excess 100%.
  • Molecular weight 618.72 g/mol and formula C37H38N4O5.
  • Supplied in small research pack sizes, e.g., 50 mg.
  • For research use only; not for human use.
Catalog No. 50-002-39468 Supplier Medchemexpress LLC Supplier No. HY12358750MG
A selective tripeptide epoxyketone inhibitor of the immunoproteasome LMP7 subunit for biochemical and cellular research. It covalently modifies proteasomal N-terminal threonine active sites and has demonstrated antitumor activity in preclinical studies.
  • Selective LMP7 (β5i) immunoproteasome inhibitor.
  • Covalently modifies N-terminal threonine active sites.
  • Reported IC50 of 22 nM against LMP7.
  • Demonstrated inhibition of multiple myeloma cell growth and induction of apoptosis.
  • High chemical purity (98.9%).
  • Enantiomeric excess 100%.
  • Molecular weight 618.72 g/mol and formula C37H38N4O5.
  • Supplied in small research pack sizes, e.g., 50 mg.
  • For research use only; not for human use.