Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenyl Antimony Dichloride, MP Biomedicals

Catalog No. ICN219546 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219546 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219546 Supplier MP Biomedicals, Inc Supplier No. 0521954605

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 594-31-0
Molecular Formula C18H15Cl2Sb
Molecular Weight (g/mol) 423.978
InChI Key PDGPVQHGCLPCES-UHFFFAOYSA-L
Synonym triphenylantimony dichloride, antimony, dichlorotriphenyl, dichlorotriphenylantimony, dichlorotriphenylstibine, antimony, triphenyldichloride, triphenylstibine dichloride, dichlorid trifenylantimonicny, dichlorid trifenylantimonicny czech, stibine, triphenyl-, dichloride 6ci, acmc-1ao41
PubChem CID 11662
IUPAC Name dichloro(triphenyl)-$l^{5}-stibane
SMILES C1=CC=C(C=C1)[Sb](C2=CC=CC=C2)(C3=CC=CC=C3)(Cl)Cl

Specifications

Quantity 10 g
Formula Weight 423.98
Chemical Name or Material Triphenyl Antimony Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.