Learn More
Medchemexpress LLC Triphenyl compound A | 21306-65-0 | 99.8% | 399.53 g/mol | C21H21NO3S2 | 5 MG
SDP
Catalog No. 5000221060
Encompass_Preferred
TPh A (Triphenyl Compound A) is a small-molecule research compound that selectively binds the nuclear protein pirin and disrupts the Bcl3-pirin complex. It is supplied as a powder for laboratory use, with high reported purity and analytical characterization suitable for cellular tool compound applications.
  • Potent pirin inhibitor with reported Ki ≈ 0.6 μM.
  • Disrupts formation of the Bcl3-pirin complex in biochemical assays.
  • High purity (99.81%) suitable for research applications.
  • Supplied as a powder for straightforward handling and aliquoting.
  • Characterized by formula C21H21NO3S2 and molecular weight 399.53 g/mol.
Catalog No. 50-002-21060 Supplier Medchemexpress LLC Supplier No. HY144545MG
TPh A (Triphenyl Compound A) is a small-molecule research compound that selectively binds the nuclear protein pirin and disrupts the Bcl3-pirin complex. It is supplied as a powder for laboratory use, with high reported purity and analytical characterization suitable for cellular tool compound applications.
  • Potent pirin inhibitor with reported Ki ≈ 0.6 μM.
  • Disrupts formation of the Bcl3-pirin complex in biochemical assays.
  • High purity (99.81%) suitable for research applications.
  • Supplied as a powder for straightforward handling and aliquoting.
  • Characterized by formula C21H21NO3S2 and molecular weight 399.53 g/mol.