Learn More
Medchemexpress LLC Triphenyl phosphate | 115-86-6 | MFCD00003031 | 99.7% | 326.3 g/mol | C18H15O4P | 100g
SDP
Catalog No. 5000267811
Encompass_Preferred
Triphenyl phosphate is an organic phosphate flame retardant Triphenyl phosphate can disrupt placental tryptophan metabolism by activating MAOA/ROS/NF B leading to abnormal neurological behavior Triphenyl phosphate promotes oxidative stress by inducing inflammatory factors like nuclear factor kappa B (NF B) interleukin-6 and tumor necrosis factor Triphenyl phosphate can also cause allergic contact dermatitis[1][2]
Catalog No. 50-002-67811 Supplier Medchemexpress LLC Supplier No. HYY1322100G
Triphenyl phosphate is an organic phosphate flame retardant Triphenyl phosphate can disrupt placental tryptophan metabolism by activating MAOA/ROS/NF B leading to abnormal neurological behavior Triphenyl phosphate promotes oxidative stress by inducing inflammatory factors like nuclear factor kappa B (NF B) interleukin-6 and tumor necrosis factor Triphenyl phosphate can also cause allergic contact dermatitis[1][2]