Learn More
Medchemexpress LLC Triphenyl(propyl)phosphonium-d5 (bromide) | 1703757-22-5 | 99.0% | 390.31 | 25 MG
SDP
Catalog No. 5000453259
Encompass_Preferred
Triphenyl(propyl)phosphonium-d5 bromide is a deuterium-labeled Triphenyl(propyl)phosphonium bromide that can be used as a tracer and an internal standard for quantitative analysis. Deuteration of drug molecules has the potential to affect their pharmacokinetic and metabolic profiles, making this compound useful in drug development research.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
Catalog No. 50-004-53259 Supplier Medchemexpress LLC Supplier No. 25MGHYW008678S
Triphenyl(propyl)phosphonium-d5 bromide is a deuterium-labeled Triphenyl(propyl)phosphonium bromide that can be used as a tracer and an internal standard for quantitative analysis. Deuteration of drug molecules has the potential to affect their pharmacokinetic and metabolic profiles, making this compound useful in drug development research.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only