Learn More
Medchemexpress LLC Triphenyl(propyl)phosphonium-d5 bromide | 1703757-22-5 | 99% | 390.31 | 100 MG
SDP
Catalog No. 5000453666
Encompass_Preferred
Triphenyl(propyl)phosphonium-d5 (bromide) is the deuterium labeled Triphenyl(propyl)phosphonium bromide. It is for research use only and not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-53666 Supplier Medchemexpress LLC Supplier No. HYW008678S100MG
Triphenyl(propyl)phosphonium-d5 (bromide) is the deuterium labeled Triphenyl(propyl)phosphonium bromide. It is for research use only and not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS