Learn More
Medchemexpress LLC Triphenyl(propyl)phosphonium-d5 bromide | 1703757-22-5 | 99% | 390.31 | 50 MG
SDP
Catalog No. 5000451169
Encompass_Preferred
Triphenyl(propyl)phosphonium-d5 (bromide) is the deuterium labeled Triphenyl(propyl)phosphonium bromide. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-51169 Supplier Medchemexpress LLC Supplier No. HYW008678S50MG
Triphenyl(propyl)phosphonium-d5 (bromide) is the deuterium labeled Triphenyl(propyl)phosphonium bromide. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS