Learn More
Medchemexpress LLC Triphenylamine-d15 | 880462-20-4 | 99.0% | 260.41 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW011998S
Encompass_Preferred
This product is a stable isotope-labeled compound, specifically a deuterium-labeled version of Triphenylamine. These types of compounds are primarily used in research applications, particularly as tracers for quantitative analysis during drug development. The incorporation of stable heavy isotopes like deuterium can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
  • For research use only.
Catalog No. 50-004-52129