Learn More
Medchemexpress LLC Triphenylamine-d15 | 880462-20-4 | 99% | 260.41 | 10 MG
SDP
Catalog No. 5000452416
Encompass_Preferred
Triphenylamine-d15 is the deuterium labeled Triphenylamine. It is for research use only and not sold to patients. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-52416 Supplier Medchemexpress LLC Supplier No. HYW011998S10MG
Triphenylamine-d15 is the deuterium labeled Triphenylamine. It is for research use only and not sold to patients. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.