Learn More
Medchemexpress LLC Triphenylamine-d15 | 880462-20-4 | 99% | 260.41 | 100 MG
SDP
Catalog No. 5000452740
Encompass_Preferred
Triphenylamine-d15 is the deuterium labeled Triphenylamine. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs. This compound is identified as a laboratory chemical used for the manufacture of substances.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
  • Identified uses: Laboratory chemicals, manufacture of substances.
  • Product is for research use only.
Catalog No. 50-004-52740 Supplier Medchemexpress LLC Supplier No. HYW011998S100MG
Triphenylamine-d15 is the deuterium labeled Triphenylamine. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs. This compound is identified as a laboratory chemical used for the manufacture of substances.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
  • Identified uses: Laboratory chemicals, manufacture of substances.
  • Product is for research use only.