Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylantimony 95.0+%, TCI America™
SDP

Catalog No. T047625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T047625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T047625G Supplier TCI America Supplier No. T047625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 603-36-1
Molecular Formula C18H15Sb
Molecular Weight (g/mol) 353.078
MDL Number MFCD00003000
InChI Key HVYVMSPIJIWUNA-UHFFFAOYSA-N
Synonym triphenylantimony, triphenylstibine, stibine, triphenyl, trifenylstibin, triphenyl stibine, trifenylstibin czech, unii-g1x1263amm, ccris 1300, triphenylantimony iii, triphenyl-stibane
PubChem CID 11777
IUPAC Name triphenylstibane
SMILES C1=CC=C(C=C1)[Sb](C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 377°C
Quantity 25 g
UN Number 3467
Formula Weight 353.08
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylantimony
EINECSNumber (3)-3436
RTECSNumber WJ1400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.