Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylarsine 97%, Thermo Scientific™

Catalog No. AC140330050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC140330050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC140330050 Supplier Thermo Scientific Chemicals Supplier No. 140330050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-32-7
Molecular Formula C18H15As
Molecular Weight (g/mol) 306.24
MDL Number MFCD00002994
InChI Key BPLUKJNHPBNVQL-UHFFFAOYSA-N
Synonym triphenylarsine, arsine, triphenyl, triphenyl arsenic, triphenylarsene, arsinetriphenyl, unii-mn8ez3fl74, mn8ez3fl74, triphenylarsenic, triphenyl arsine, triphenyl-arsine
PubChem CID 11773
IUPAC Name triphenylarsane
SMILES C1=CC=C(C=C1)[As](C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 57°C to 62°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula (C6H5)3As
Quantity 5 g
Beilstein 16, 828
Fieser 17,262
Formula Weight 306.23
Percent Purity ≥96.0%
Physical Form Crystalline Solid
Chemical Name or Material Triphenylarsine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.