Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylbismuth Dichloride 98.0+%, TCI America™
SDP

Catalog No. T18375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18375G Supplier TCI America Supplier No. T18375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 594-30-9
Molecular Formula C18H15BiCl2
Molecular Weight (g/mol) 511.198
MDL Number MFCD00015725
InChI Key QWMMHFXTVYRMTO-UHFFFAOYSA-L
Synonym Dichlorotriphenylbismuth
PubChem CID 299728
ChEBI CHEBI:37199
IUPAC Name dichloro(triphenyl)bismuth
SMILES C1=CC=C(C=C1)[Bi](C2=CC=CC=C2)(C3=CC=CC=C3)(Cl)Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 511.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylbismuth Dichloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.