Learn More
Strem, An Ascensus Company CAS# 960-71-4. 5g. Triphenylborane, min. 95%. MFCD00003007
SDP
Supplier:  Strem, An Ascensus Company 9305435G
Encompass
CAS# 960-71-4. 5g. Triphenylborane, min. 95%. MFCD00003007. Molecular Weight: 242.13. Molecular Formula: B(C6H5)3. Color/form: off-white pwdr. Strem# 93-0543. http://www.strem.com/catalog/v/93-0543/
Catalog No. 50-901-13743