Learn More
Medchemexpress LLC Triphenylene-d12 | 17777-56-9 | 98.0% | 240.36 | 10 MG
SDP
Catalog No. 5000365095
Encompass_Preferred
Triphenylene-d12 is a deuterium labeled triphenylene, designed for research use. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of compounds. This substance is intended for laboratory applications, not for patient use.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS techniques
Catalog No. 50-003-65095 Supplier Medchemexpress LLC Supplier No. 10MGHYW017708S
Triphenylene-d12 is a deuterium labeled triphenylene, designed for research use. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of compounds. This substance is intended for laboratory applications, not for patient use.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS techniques