Learn More
Medchemexpress LLC Triphenylene-d12 | 17777-56-9 | 98% | 240.36 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHYW017708S
Encompass_Preferred
Triphenylene-d12 is the deuterium labeled triphenylene, intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis, suitable for methods like NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-64957