Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triphenylmethane

Catalog No. 10130325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-130-325G 25 g
10-130-3500G 500 g
10-130-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-130-325G Supplier Sigma Aldrich Supplier No. 10130325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triphenylmethane

Specifications

CAS 519-73-3
Melting Point 92°C to 94°C (lit.)
Density 1.014 g/mL (at 25°C (literature))
Boiling Point 358°C to 359°C (754 mmHg)
Molecular Formula C19H16
Linear Formula (C6H5)3 CH
MDL Number MFCD00004763
Quantity 25 g
Molecular Weight (g/mol) 244.33
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.