Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylmethane, MP Biomedicals

Catalog No. ICN219570 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219570 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219570 Supplier MP Biomedicals, Inc Supplier No. 0521957010

Chemical Identifiers

CAS 519-73-3
Molecular Formula C19H16
Molecular Weight (g/mol) 244.34
MDL Number MFCD00004763
InChI Key AAAQKTZKLRYKHR-UHFFFAOYSA-N
Synonym triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane?
PubChem CID 10614
ChEBI CHEBI:76212
IUPAC Name (diphenylmethyl)benzene
SMILES C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 10 g
Formula Weight 244.34
Chemical Name or Material Triphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.