Learn More
Medchemexpress LLC Triphenylmethanol | 76-84-6 | 99.84% | 260.34 | 1 ML
SDP
Catalog No. 5000453814
Encompass_Preferred
Triphenylmethanol is a specific clathrate host for methanol and dimethyl sulphoxide. It also inhibits the neoplastic transformation of cells.
  • Specific clathrate host for methanol and dimethyl sulphoxide.
  • Inhibits neoplastic transformation of cells.
Catalog No. 50-004-53814 Supplier Medchemexpress LLC Supplier No. HYW01178810MM/1ML
Add to Cart
Add to Cart
Triphenylmethanol is a specific clathrate host for methanol and dimethyl sulphoxide. It also inhibits the neoplastic transformation of cells.
  • Specific clathrate host for methanol and dimethyl sulphoxide.
  • Inhibits neoplastic transformation of cells.