Learn More
Medchemexpress LLC Triphenylmethanol | 76-84-6 | 99.96% | 260.34 | 500 G
SDP
Catalog No. 5000453786
Encompass_Preferred
Triphenylmethanol is a specific clathrate host for methanol and dimethyl sulphoxide. It also inhibits the neoplastic transformation of cells.
  • Inhibits neoplastic transformation of cells
  • Specific clathrate host for methanol
  • Specific clathrate host for dimethyl sulphoxide
Catalog No. 50-004-53786 Supplier Medchemexpress LLC Supplier No. HYW011788500G
Triphenylmethanol is a specific clathrate host for methanol and dimethyl sulphoxide. It also inhibits the neoplastic transformation of cells.
  • Inhibits neoplastic transformation of cells
  • Specific clathrate host for methanol
  • Specific clathrate host for dimethyl sulphoxide