Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylmethyl Chloride 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
76-83-5
C19H15Cl
278.77
MFCD00000813
JBWKIWSBJXDJDT-UHFFFAOYSA-N
trityl chloride,triphenylmethyl chloride,triphenylchloromethane,chlorotriphenylmethane,chloromethanetriyl tribenzene,triphenylmethylchloride,methane, chlorotriphenyl,triphenyl chloromethane,chlorodiphenylmethyl benzene,benzene, 1,1',1-chloromethylidyne tris
6456
[chloro(diphenyl)methyl]benzene
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
76-83-5
C19H15Cl
278.77
MFCD00000813
JBWKIWSBJXDJDT-UHFFFAOYSA-N
trityl chloride,triphenylmethyl chloride,triphenylchloromethane,chlorotriphenylmethane,chloromethanetriyl tribenzene,triphenylmethylchloride,methane, chlorotriphenyl,triphenyl chloromethane,chlorodiphenylmethyl benzene,benzene, 1,1',1-chloromethylidyne tris
6456
[chloro(diphenyl)methyl]benzene
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
Quantity 100g
Packaging Plastic bottle
Infrared Spectrum Authentic
Color Beige-Yellow or White
Melting Point 110.0°C to 113.0°C
Boiling Point 230.0°C to 235.0°C (20.0mmHg)
Assay Percent Range 98%
Linear Formula (C6H5)3CCl
Beilstein 05, 700
Fieser 01,1254; 02,453; 04,565; 05,741; 06,656; 15,359
Merck Index 15, 9944
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene, dichloromethane and dmf
Formula Weight 278.77
Physical Form Powder or Crystals
Percent Purity 98%
Chemical Name or Material Triphenylmethyl chloride, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-986-4

RTECSNumber : PA6450000

TSCA : TSCA